Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134562.1 | NP_001128034.1 | 13 | 2091 | ||
XM_006241059.2 | XP_006241121.1 | 12 | 2571 |