Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108084.1 | NP_001101554.1 | 2 | 830 | ||
XM_006241255.2 | XP_006241317.1 | 2 | 810 | |||
XM_006241261.2 | XP_006241323.1 | 3 | 1005 | |||
XM_006241263.2 | XP_006241325.1 | 2 | 1653 |