Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108099.1 | NP_001101569.1 | 11 | 1319 | ||
XM_006241382.1 | XP_006241444.1 | 12 | 1684 | |||
XM_006241383.1 | XP_006241445.1 | 12 | 1248 | |||
XM_008765396.1 | XP_008763618.1 | 13 | 1756 | |||
XM_008765397.1 | XP_008763619.1 | 13 | 1320 | |||
XM_008765399.1 | XP_008763621.1 | 9 | 1066 |