Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Twf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008521.1 | NP_001008521.1 | 7 | 741 | ||
GenBank mRNA | BC086536.1 | - | 7 | 741 | 7106678 |