Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014087.1 | NP_001014109.2 | 7 | 856 | ||
XM_008765933.1 | XP_008764155.1 | 7 | 1039 | |||
XM_008765934.1 | XP_008764156.1 | 7 | 972 | |||
XM_008765935.1 | XP_008764157.1 | 6 | 1081 | |||
XM_008765936.1 | XP_008764158.1 | 7 | 1039 | |||
XM_008765937.1 | XP_008764159.1 | 7 | 1037 | |||
XM_008765938.1 | XP_008764160.1 | 7 | 1037 | |||
GenBank mRNA | BC085333.1 | - | 7 | 856 | 7114511 |