Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elof1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126098.1 | NP_001119570.1 | 2 | 326 | ||
XM_008765987.1 | XP_008764209.1 | 2 | 115 | |||
XM_008765988.1 | XP_008764210.1 | 2 | 152 | |||
XM_008765989.1 | XP_008764211.1 | 2 | 148 |