Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207610.1 | NP_997493.1 | 15, 16 | 2559 | ||
XM_006242938.2 | XP_006243000.1 | 16, 17 | 2773 | |||
GenBank mRNA | AY325198.1 | - | 15, 16 | 2511 | ||
BC061761.1 | - | 15, 16 | 2559 | 5621078 |