Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fdxacb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108145.1 | NP_001101615.1 | 4 | 1680 | ||
XM_006243031.2 | XP_006243093.1 | 5 | 2204 |