Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766393.1 | XP_008764615.1 | 23 | 4210 | ||
XM_008766394.1 | XP_008764616.1 | 23 | 4376 | |||
XM_008766395.1 | XP_008764617.1 | 23 | 4210 |