Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766402.1 | XP_008764624.1 | 8 | 946 | ||
XM_008766403.1 | XP_008764625.1 | 8 | 2984 | |||
XM_008766404.1 | XP_008764626.1 | 8 | 946 | |||
XM_008766405.1 | XP_008764627.1 | 8 | 946 |