Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acad11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108181.1 | NP_001101651.1 | 1 | 197 | ||
GenBank mRNA | BC167762.1 | - | 1 | 202 | 7443858 |