Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108197.1 | NP_001101667.1 | 7 | 721 | ||
XM_008766860.1 | XP_008765082.1 | 7 | 722 | |||
XM_008766861.1 | XP_008765083.1 | 7 | 722 | |||
XM_008766862.1 | XP_008765084.1 | 7 | 722 | |||
GenBank mRNA | FQ211123.1 | - | 7 | 718 |