Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102548228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244873.1 | XP_006244935.1 | 6 | 1515 | ||
XM_006244874.2 | XP_006244936.1 | 6 | 1088 | |||
XM_006244875.2 | XP_006244937.1 | 6 | 1303 | |||
XM_006244876.1 | XP_006244938.1 | 6 | 1235 | |||
XM_006244879.1 | XP_006244941.1 | 6 | 1423 | |||
XM_006244881.2 | XP_006244943.1 | 6 | 1212 | |||
XM_008767062.1 | XP_008765284.1 | 6 | 1041 | |||
XM_008767063.1 | XP_008765285.1 | 5 | 476 |