Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsfy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012132.1 | NP_001012132.1 | 1 | 995 | ||
GenBank mRNA | BC083618.1 | - | 1 | 995 | 7133881 |