Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102547271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256847.2 | XP_006256909.1 | 2 | 216 |
Gene Symbol: Msl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014111.1 | NP_001014133.1 | 5 | 504 | ||
XM_006256835.2 | XP_006256897.2 | 6 | 890 | |||
XM_006256836.2 | XP_006256898.2 | 6 | 893 | |||
XM_006256839.2 | XP_006256901.2 | 6 | 890 | |||
XM_008773169.1 | XP_008771391.1 | 6 | 893 | |||
GenBank mRNA | BC086548.1 | - | 5 | 504 | 7301795 |