Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ston1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182954.2 | NP_891999.2 | 2 | 1082 | ||
XM_006239743.2 | XP_006239805.1 | 2 | 853 | |||
XM_006239744.2 | XP_006239806.1 | 2 | 1949 | |||
XM_008764479.1 | XP_008762701.1 | 3 | 1170 | |||
XM_008764480.1 | XP_008762702.1 | 2 | 1638 | |||
XM_008764481.1 | XP_008762703.1 | 2 | 833 | |||
GenBank mRNA | AY300793.1 | - | 1 | 800 |