Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108274.1 | NP_001101744.1 | 6 | 998 | ||
XM_006246291.2 | XP_006246353.1 | 6 | 1025 | |||
XM_006246294.2 | XP_006246356.1 | 6 | 1025 | |||
XM_006246295.2 | XP_006246357.1 | 6 | 1025 | |||
XM_008767690.1 | XP_008765912.1 | 6 | 1026 | |||
GenBank mRNA | BC166483.1 | - | 6 | 934 | 7099086 |