Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tada2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012141.1 | NP_001012141.1 | 6 | 535 | ||
XM_006247041.2 | XP_006247103.1 | 5 | 498 | |||
XM_006247042.2 | XP_006247104.1 | 5 | 444 | |||
GenBank mRNA | BC079084.1 | - | 6 | 535 | 7123007 | |
EF121983.1 | - | 5 | 374 | |||
EF121984.1 | - | 5 | 374 | |||
EF121985.1 | - | 5 | 374 |