Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggnbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004273.1 | NP_001004273.1 | 9 | 1506 | ||
XM_006247047.2 | XP_006247109.1 | 11 | 1720 | |||
XM_006247048.2 | XP_006247110.1 | 11 | 1713 | |||
XM_006247049.2 | XP_006247111.1 | 11 | 1717 | |||
XM_006247050.2 | XP_006247112.1 | 11 | 1710 | |||
XM_006247051.2 | XP_006247113.1 | 10 | 1592 | |||
XM_006247052.2 | XP_006247114.1 | 10 | 1586 | |||
XM_006247053.2 | XP_006247115.1 | 10 | 1589 | |||
XM_006247054.2 | XP_006247116.1 | 10 | 1582 | |||
XM_008768070.1 | XP_008766292.1 | 11 | 1694 | |||
XM_008768071.1 | XP_008766293.1 | 11 | 1664 | |||
GenBank mRNA | AY633742.1 | - | 9 | 1506 | ||
FQ220883.1 | - | 7 | 884 | |||
FQ227019.1 | - | 4 | 509 | |||
FQ234201.1 | - | 7 | 973 | |||
FQ234847.1 | - | 6 | 832 |