Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med13l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249420.2 | XP_006249482.1 | 18 | 4128 | ||
XM_006249421.2 | XP_006249483.1 | 19 | 4505 | |||
XM_006249422.2 | XP_006249484.1 | 19 | 4505 | |||
GenBank mRNA | GU566026.1 | - | 19 | 4493 |