Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1308117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134571.2 | NP_001128043.2 | 6 | 980 | ||
XM_006252278.2 | XP_006252340.1 | 6 | 3072 | |||
XM_008770829.1 | XP_008769051.1 | 5 | 1119 | |||
GenBank mRNA | CK363611.1 | - | 2 | 439 |