Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082409.1 | NP_001075878.1 | 1, 2 | 115 | ||
XM_008771539.1 | XP_008769761.1 | 1, 2 | 160 | |||
XM_008771540.1 | XP_008769762.1 | 1, 2 | 160 | |||
XM_008771541.1 | XP_008769763.1 | 1, 2 | 160 | |||
GenBank mRNA | BC167744.1 | - | 1, 2 | 104 | 7099140 |