Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3galnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144851.1 | NP_001138323.1 | 12 | 2976 | ||
XM_006254007.2 | XP_006254069.1 | 11 | 2817 | |||
XM_006254008.1 | XP_006254070.1 | 12 | 2614 | |||
XM_008771701.1 | XP_008769923.1 | 12 | 2909 | |||
XM_008771703.1 | XP_008769925.1 | 11 | 2908 | |||
GenBank mRNA | FQ212048.1 | - | 9 | 2416 |
Gene Symbol: Tbce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012161.1 | NP_001012161.1 | 17 | 1578 | ||
GenBank mRNA | BC089833.1 | - | 17 | 1578 | 7325371 | |
FQ234296.1 | - | 9 | 703 |