Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012161.1 | NP_001012161.1 | 16 | 1502 | ||
XM_006254015.2 | XP_006254077.1 | 16 | 1558 | |||
XM_008771736.1 | XP_008769958.1 | 16 | 1474 | |||
GenBank mRNA | BC089833.1 | - | 16 | 1502 | 7325371 | |
FQ234296.1 | - | 8 | 627 |
Gene Symbol: B3galnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144851.1 | NP_001138323.1 | 12 | 3228 | ||
XM_006254007.2 | XP_006254069.1 | 11 | 3069 | |||
XM_006254008.1 | XP_006254070.1 | 12 | 2866 | |||
XM_008771701.1 | XP_008769923.1 | 12 | 3161 | |||
XM_008771703.1 | XP_008769925.1 | 11 | 3160 | |||
GenBank mRNA | FQ212048.1 | - | 9 | 2668 |