Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254677.2 | XP_006254739.1 | 13 | 2349 | ||
XM_006254678.2 | XP_006254740.1 | 12 | 2184 | |||
XM_006254679.2 | XP_006254741.1 | 12 | 2184 | |||
XM_006254680.2 | XP_006254742.1 | 12 | 2055 | |||
XM_008772179.1 | XP_008770401.1 | 13 | 2349 |