Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199404.1 | NP_955436.1 | 9 | 1157 | ||
XM_006255273.2 | XP_006255335.1 | 9 | 1238 | |||
XM_006255274.2 | XP_006255336.1 | 9 | 1234 | |||
XM_006255275.2 | XP_006255337.1 | 9 | 1204 | |||
GenBank mRNA | BC061819.1 | - | 9 | 1157 | 5599365 |