Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipcef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170799.1 | NP_001164270.1 | 9 | 1271 | ||
GenBank mRNA | AJ536192.1 | - | 9 | 1271 |