Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm6sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108490.1 | NP_001101960.1 | 8, 9 | 834 | ||
XM_006229441.2 | XP_006229503.1 | 7, 8 | 716 | |||
XM_008759543.1 | XP_008757765.1 | 6, 7 | 1096 | |||
XM_008759544.1 | XP_008757766.1 | 7, 8 | 688 |