Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212508.2 | NP_997673.1 | 3, 4 | 562 | ||
GenBank mRNA | BP504258.1 | - | 3, 4 | 568 |