Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Derl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109577.1 | NP_001103047.2 | 6, 7 | 640 | ||
XM_006256325.2 | XP_006256387.1 | 6, 7 | 2029 | |||
XM_006256326.2 | XP_006256388.1 | 6, 7 | 2026 | |||
XM_008772914.1 | XP_008771136.1 | 3, 4 | 1246 | |||
XM_008772915.1 | XP_008771137.1 | 3, 4 | 1246 | |||
XM_008772916.1 | XP_008771138.1 | 3, 4 | 1246 | |||
XM_008772917.1 | XP_008771139.1 | 3, 4 | 1246 | |||
XM_008772918.1 | XP_008771140.1 | 5, 6 | 2443 | |||
XM_008772920.1 | XP_008771142.1 | 5, 6 | 2440 | |||
XM_008772921.1 | XP_008771143.1 | 4, 5 | 1135 | |||
XM_008772922.1 | XP_008771144.1 | 4, 5 | 2378 | |||
XM_008772923.1 | XP_008771145.1 | 4, 5 | 835 | |||
XM_008772924.1 | XP_008771146.1 | 4, 5 | 835 | |||
GenBank mRNA | BC167034.1 | - | 6, 7 | 597 |
Gene Symbol: LOC103694875 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773829.1 | XP_008772051.1 | 3, 4 | 1246 | ||
XM_008773830.1 | XP_008772052.1 | 3, 4 | 1246 | |||
XM_008773831.1 | XP_008772053.1 | 3, 4 | 1246 | |||
XM_008773832.1 | XP_008772054.1 | 3, 4 | 1246 | |||
XM_008773833.1 | XP_008772055.1 | 5, 6 | 2443 | |||
XM_008773835.1 | XP_008772057.1 | 5, 6 | 2440 | |||
XM_008773836.1 | XP_008772058.1 | 4, 5 | 1135 | |||
XM_008773837.1 | XP_008772059.1 | 4, 5 | 2378 | |||
XM_008773838.1 | XP_008772060.1 | 4, 5 | 835 | |||
XM_008773839.1 | XP_008772061.1 | 4, 5 | 835 | |||
XM_008773840.1 | XP_008772062.1 | 6, 7 | 2028 | |||
XM_008773841.1 | XP_008772063.1 | 6, 7 | 2029 | |||
XM_008773842.1 | XP_008772064.1 | 6, 7 | 2026 |