Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085353.1 | NP_001078822.1 | 6 | 1005 | ||
NM_199408.2 | NP_955440.1 | 6 | 1011 | |||
GenBank mRNA | AY569010.1 | - | 6 | 1011 | ||
BC062391.1 | - | 6 | 1005 | 5621215 |