Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1560470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233652.2 | XP_006233714.1 | 2 | 1763 | ||
XM_006233653.2 | XP_006233715.1 | 3 | 1678 | |||
XM_008761615.1 | XP_008759837.1 | 2 | 3098 | |||
GenBank mRNA | FQ211897.1 | - | 3 | 408 |