Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108577.2 | NP_001102047.1 | 4 | 495 | ||
GenBank mRNA | BC166861.1 | - | 4 | 495 | 9101723 | |
FQ223957.1 | - | 5 | 650 |