Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077429.1 | NP_001070897.1 | 9 | 751 | ||
XM_006234377.2 | XP_006234439.1 | 9 | 833 | |||
XM_006234378.2 | XP_006234440.1 | 10 | 854 | |||
XM_006234379.2 | XP_006234441.1 | 9 | 881 | |||
XM_006234380.2 | XP_006234442.1 | 9 | 953 | |||
XM_006234381.2 | XP_006234443.1 | 9 | 1046 | |||
XM_006234382.2 | XP_006234444.1 | 10 | 974 | |||
XM_006234385.2 | XP_006234447.1 | 9 | 835 | |||
XM_008761909.1 | XP_008760131.1 | 9 | 1930 | |||
XM_008761910.1 | XP_008760132.1 | 9 | 835 | |||
GenBank mRNA | BC126100.1 | - | 9 | 751 | 7132654 | |
FQ218047.1 | - | 8 | 630 |