Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caprin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012185.2 | NP_001012185.2 | 5, 6 | 675 | ||
XM_006234652.2 | XP_006234714.1 | 6, 7 | 730 | |||
GenBank mRNA | AB373991.1 | - | 4, 5 | 590 | ||
AY155572.1 | - | 4, 5 | 531 | |||
BC087123.1 | - | 4, 5 | 545 |