Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpped2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198778.2 | NP_942073.2 | 5 | 774 | ||
XM_006234680.2 | XP_006234742.1 | 5 | 2149 | |||
XM_006234681.2 | XP_006234743.1 | 5 | 851 | |||
XM_006234682.2 | XP_006234744.1 | 5 | 938 | |||
XM_006234683.2 | XP_006234745.1 | 5 | 853 | |||
XM_008762104.1 | XP_008760326.1 | 5 | 1573 | |||
XM_008762105.1 | XP_008760327.1 | 4 | 1833 | |||
GenBank mRNA | AY093422.1 | - | 5 | 824 | ||
BC161828.1 | - | 5 | 856 | 9033590 | ||
EV768250.1 | - | 3 | 409 |