Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109630.1 | NP_001103100.1 | 16 | 2057 | ||
XM_006234710.2 | XP_006234772.1 | 15 | 3032 | |||
XM_006234711.2 | XP_006234773.1 | 16 | 2614 |