Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277235.1 | NP_001264164.1 | 2 | 263 | ||
XM_006235126.2 | XP_006235188.1 | 3 | 456 | |||
XM_006235127.2 | XP_006235189.1 | 3 | 445 | |||
GenBank mRNA | FQ211753.1 | - | 2 | 263 |