Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191857.1 | NP_001178786.1 | 13 | 1301 | ||
XM_006235772.2 | XP_006235834.1 | 13 | 1357 | |||
XM_006235773.2 | XP_006235835.1 | 13 | 1766 | |||
XM_006235774.2 | XP_006235836.1 | 13 | 1357 | |||
XM_006235775.2 | XP_006235837.1 | 13 | 1357 | |||
XM_006235776.2 | XP_006235838.1 | 13 | 1357 | |||
XM_006235778.2 | XP_006235840.1 | 6 | 896 |