Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cspp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191864.1 | NP_001178793.1 | 30 | 3507 | ||
XM_006237763.2 | XP_006237825.1 | 31 | 3803 | |||
XM_006237767.2 | XP_006237829.1 | 30 | 3650 | |||
XM_006237774.2 | XP_006237836.1 | 26 | 3239 | |||
XM_006237775.2 | XP_006237837.1 | 31 | 3800 | |||
XM_006237776.2 | XP_006237838.1 | 30 | 3776 | |||
XM_006237777.2 | XP_006237839.1 | 29 | 3695 | |||
XM_006237778.2 | XP_006237840.1 | 30 | 3647 | |||
XM_008763498.1 | XP_008761720.1 | 31 | 3803 | |||
XM_008763499.1 | XP_008761721.1 | 31 | 3791 | |||
XM_008763500.1 | XP_008761722.1 | 30 | 3779 | |||
XM_008763501.1 | XP_008761723.1 | 29 | 3698 | |||
XM_008763503.1 | XP_008761725.1 | 28 | 3599 | |||
XM_008763504.1 | XP_008761726.1 | 31 | 3714 | |||
XM_008763505.1 | XP_008761727.1 | 30 | 3599 | |||
XM_008763506.1 | XP_008761728.1 | 31 | 4290 | |||
GenBank mRNA | BC166546.1 | - | 12 | 1325 |