Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108682.1 | NP_001102152.1 | 1 | 2112 | ||
XM_008764148.1 | XP_008762370.1 | 1 | 2666 |