Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012191.1 | NP_001012191.1 | 7 | 955 | ||
XM_006239816.2 | XP_006239878.1 | 7 | 894 | |||
XM_006239817.2 | XP_006239879.1 | 7 | 893 | |||
XM_006239818.2 | XP_006239880.1 | 7 | 893 | |||
XM_006239819.2 | XP_006239881.1 | 7 | 894 | |||
XM_006239822.2 | XP_006239884.1 | 7 | 893 | |||
GenBank mRNA | BC081889.1 | - | 7 | 955 | 7109493 |