Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108708.1 | NP_001102178.1 | 17 | 2002 | ||
XM_006240022.2 | XP_006240084.1 | 20 | 2724 | |||
XM_006240023.2 | XP_006240085.1 | 20 | 2689 |