Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hectd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240086.1 | XP_006240148.1 | 12 | 2110 | ||
XM_006240087.1 | XP_006240149.1 | 12 | 2110 | |||
XM_006240088.1 | XP_006240150.1 | 12 | 2110 | |||
XM_006240089.1 | XP_006240151.1 | 12 | 2110 |