Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcpkmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764728.1 | XP_008762950.1 | 5 | 673 | ||
GenBank mRNA | FQ209696.1 | - | 662 |