Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5dc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134887.1 | NP_001128359.1 | 14 | 1584 | ||
XM_006241203.2 | XP_006241265.1 | 14 | 1584 | |||
XM_006241204.2 | XP_006241266.1 | 14 | 1586 |