Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130561.2 | NP_001124033.1 | 10 | 1747 | ||
XM_006241614.2 | XP_006241676.1 | 10 | 1843 | |||
GenBank mRNA | BC169014.1 | - | 10 | 1747 | 9091867 | |
FQ213462.1 | - | 5 | 694 | |||
FQ222235.1 | - | 2 | 397 |