Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbfox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079895.1 | NP_001073364.1 | 10, 11 | 1558 | ||
XM_006241834.2 | XP_006241896.1 | 11, 12 | 1610 | |||
XM_006241836.2 | XP_006241898.1 | 11, 12 | 1598 | |||
XM_006241837.2 | XP_006241899.1 | 11, 12 | 1595 | |||
GenBank mRNA | BC128766.1 | - | 10, 11 | 1558 | 7123798 |