Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Micall1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729832.4 | XP_002729878.2 | 6 | 1240 | ||
XM_006242043.2 | XP_006242105.1 | 6 | 1859 | |||
XM_006242044.2 | XP_006242106.1 | 3 | 963 | |||
XM_008765803.1 | XP_008764025.1 | 6 | 1442 | |||
XM_008765804.1 | XP_008764026.1 | 6 | 1574 |