Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009713.2 | NP_001009713.1 | 7, 8 | 1011 | ||
GenBank mRNA | AY800277.1 | - | 7, 8 | 1010 | ||
BC097482.1 | - | 7, 8 | 1011 | 7459136 |